D-Prolinamide ≥98.0%

Supplier: TCI America

Synonyms: H-D-Pro-NH₂, (R)-2-Pyrrolidinecarboxamide, H-D-Pro-NH2

P2083-1G P2083-5G
TCP2083-1GEA 100.8 CAD
TCP2083-1G TCP2083-5G
D-Prolinamide ≥98.0%
D-Prolinamide

CAS Number: 62937-45-5
MDL Number: MFCD00153457
Molecular Formula: C5H10N2O
Molecular Weight: 114.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 102
Specific rotation [a]20/D: 109 deg (C=2, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Amino Acid Amides
Other Product Names / Synonyms: (R)-2-Pyrrolidinecarboxamide

Formula: C₅H₁₀N₂O
Melting Pt: 102 °C
MDL Number: MFCD00153457
CAS Number: 62937-45-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb46fc89922c8de12a875d1c1f591c04cc0ad3e4&vid=eb46fc89922c8de12a875d1c1f591c04cc0ad3e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR