Psoralen ≥99.0%
Supplier: TCI America
Synonyms:
7H-Furo[3,2-g]chromen-7-one, Ficusin
P2077-100MG
P2077-20MG
TCP2077-100MGEA
344.27
CAD
TCP2077-100MG
TCP2077-20MG
Psoralen ≥99.0%
Psoralen
CAS Number: 66-97-7
MDL Number: MFCD00010520
Molecular Formula: C11H6O3
Molecular Weight: 186.17
Purity/Analysis Method: >99.0% (GC)
Melting point (°C): 163
Storage Temperature: 0-10°C
Product Categories: Bioscience, Reagents for Inflammation & Immunity Disorder Models, Phenylpropanoids
Other Product Names / Synonyms: 7H-Furo[3,2-g]benzopyran-7-one, Furo[3,2-g]coumarin
Formula:
C₁₁H₆O₃ Melting Pt: 163 °C |
MDL Number:
MFCD00010520 CAS Number: 66-97-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...