3-(1-Piperazinyl)-1,2-benzisothiazole ≥98.0%
Supplier: TCI America
Synonyms:
Ziprasidone EP Impurity A
P2064-1G
P2064-5G
TCP2064-1GEA
96.38
CAD
TCP2064-1G
TCP2064-5G
3-(1-Piperazinyl)-1,2-benzisothiazole ≥98.0%
3-(1-Piperazinyl)-1,2-benzisothiazole
CAS Number: 87691-87-0
MDL Number: MFCD04117970
Molecular Formula: C11H13N3S
Molecular Weight: 219.31
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 91
Storage Temperature: 0-10°C
Formula:
C₁₁H₁₃N₃S Melting Pt: 91 °C |
MDL Number:
MFCD04117970 CAS Number: 87691-87-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...