3-Phenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3(5)-Phenyl-1H-pyrazole, 3-Phenylpyrazole

P2063-1G P2063-5G
TCP2063-1GEA 150.1 CAD
TCP2063-1G TCP2063-5G
3-Phenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
3-Phenyl-1H-pyrazole

CAS Number: 2458-26-6
MDL Number: MFCD00159654
Molecular Formula: C9H8N2
Molecular Weight: 144.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Pale Yellow
Boiling point (°C): 314
Melting point (°C): 78

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 314 °C
Melting Pt: 78 °C
MDL Number: MFCD00159654
CAS Number: 2458-26-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e327d388bb4fe33ae4158d30052089675e8899bb&vid=e327d388bb4fe33ae4158d30052089675e8899bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR