Pramiracetam hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P2061-1G P2061-5G
TCP2061-1GEA 61.62 CAD
TCP2061-1G TCP2061-5G
Pramiracetam hydrate ≥98.0% (by HPLC, titration analysis)
Pramiracetam hydrate

CAS Number: 68497-62-1
MDL Number: MFCD00867219
Molecular Formula: C14H27N3O2
Molecular Weight: 269.39
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Boiling point (°C): 164
Melting point (°C): 47

Formula: C₁₄H₂₇N₃O₂
Boiling Pt: 164 °C
Melting Pt: 47 °C
MDL Number: MFCD00867219
CAS Number: 68497-62-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14990d40fb6974d3319bfb0ff76444627b6b58ab&vid=14990d40fb6974d3319bfb0ff76444627b6b58ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR