2-Phenylquinoline ≥98.0%

Supplier: TCI America

P2057-1G P2057-5G
TCP2057-1GEA 97.96 CAD
TCP2057-1G TCP2057-5G
2-Phenylquinoline ≥98.0%
2-Phenylquinoline

CAS Number: 612-96-4
MDL Number: MFCD00011568
Molecular Formula: C15H11N
Molecular Weight: 205.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 363
Melting point (°C): 83
Lambda max.: 258 nm

Formula: C₁₅H₁₁N
Boiling Pt: 363 °C
Melting Pt: 83 °C
MDL Number: MFCD00011568
CAS Number: 612-96-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a64ee5790d3917214e81c7375473a0392e01eb04&vid=a64ee5790d3917214e81c7375473a0392e01eb04&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR