1-Phenyl-1,2,3,4-tetrahydroisoquinoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P2056-1G P2056-5G
TCP2056-1GEA 66.36 CAD
TCP2056-1G TCP2056-5G
1-Phenyl-1,2,3,4-tetrahydroisoquinoline ≥98.0% (by GC, titration analysis)
1-Phenyl-1,2,3,4-tetrahydroisoquinoline

CAS Number: 22990-19-8
MDL Number: MFCD02179241
Molecular Formula: C15H15N
Molecular Weight: 209.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 100
Lambda max.: 265 nm (EtOH)


Other Product Names / Synonyms: 1,2,3,4-Tetrahydro-1-phenylisoquinoline

Formula: C₁₅H₁₅N
Melting Pt: 100 °C
MDL Number: MFCD02179241
CAS Number: 22990-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d69a5482361bad38bd41496b35dfb3003fcab84b&vid=d69a5482361bad38bd41496b35dfb3003fcab84b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR