Pararosaniline hydrochloride ≥95.0% (by total nitrogen basis) for biochemical research
Supplier: TCI America
Synonyms:
Basic Parafuchsin, 4,4'-(4-Iminocyclohexa-2,5-dienylidenemethylene)dianiline hydrochloride, [4-[Bis(4-aminophenyl)methylidene]-1-cyclohexa-2,5-dienylidene]azanium chloride, Basic Fuchsin hydrochloride, Parafuchsin hydrochloride, Pararosaniline chloride, Basic Red 9, 4,4'-((4-Iminocyclohexa-2,5-dien-1-ylidene)methylene)dianiline hydrochloride, Paramagenta hydrochloride
[for Biochemical Research]
CAS Number: 569-61-9
MDL Number: MFCD00001657
Molecular Formula: C19H17N3
Molecular Weight: 323.82
Purity/Analysis Method:
Product Categories: Bioscience, Glucuronidase Substrates
Other Product Names / Synonyms: Parafuchsin, Basic Red 9
Formula:
C₁₉H₁₈ClN₃ Melting Pt: 270 °C |
MDL Number:
MFCD00001657 CAS Number: 569-61-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...