Prilocaine hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

P2038-1G P2038-5G
TCP2038-1GEA 80.58 CAD
TCP2038-1G TCP2038-5G
Prilocaine hydrochloride ≥98.0% (by HPLC)
Prilocaine hydrochloride

CAS Number: 1786-81-8
MDL Number: MFCD00079279
Molecular Formula: C13H20N2O
Molecular Weight: 256.77
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 166


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 2-(Propylamino)-N-(o-tolyl)propionamide Hydrochloride

Formula: C₁₃H₂₀N₂O
Melting Pt: 166 °C
MDL Number: MFCD00079279
CAS Number: 1786-81-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a707f1fe0bf9c75341b933c8c37f9516dbea2604&vid=a707f1fe0bf9c75341b933c8c37f9516dbea2604&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR