Polyethylene glycol 2,000

Supplier: TCI America

P2034-25G P2034-500G
TCP2034-25GEA 49.6 CAD
TCP2034-25G TCP2034-500G
Polyethylene glycol 2,000
Polyethylene glycol 2,000

CAS Number: 25322-68-3
MDL Number: MFCD01779611
Form: Crystal
Color: White
Melting point (°C): 55
Flash Point (°C): 229


Other Product Names / Synonyms: PEG 2,000

Formula: H(OCH₂CH₂)nOH
Melting Pt: 55 °C
Flash Pt: 229 °C
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR