1-Phenylimidazole ≥98.0% (by GC)

Supplier: TCI America

P2030-1G P2030-5G
TCP2030-1GEA 95.91 CAD
TCP2030-1G TCP2030-5G
1-Phenylimidazole ≥98.0% (by GC)
1-Phenylimidazole

CAS Number: 7164-98-9
MDL Number: MFCD00041204
Molecular Formula: C9H8N2
Molecular Weight: 144.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 142
Melting point (°C): 13
Flash Point (°C): 113
Specific Gravity (20/20): 1.14

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 142 °C
Melting Pt: 13 °C
Density: 1.14 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00041204
CAS Number: 7164-98-9
UN: 2384
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116dc28ed0411c5e1db6feacd41e335fa823b04e&vid=116dc28ed0411c5e1db6feacd41e335fa823b04e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR