5-Phenyl-3-isoxazolecarboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P2020-1G P2020-5G
TCP2020-1GEA 150.42 CAD
TCP2020-1G TCP2020-5G
5-Phenyl-3-isoxazolecarboxylic acid ≥98.0% (by HPLC, titration analysis)
5-Phenyl-3-isoxazolecarboxylic acid

CAS Number: 14441-90-8
MDL Number: MFCD00464221
Molecular Formula: C10H7NO3
Molecular Weight: 189.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 160

Formula: C₁₀H₇NO₃
Melting Pt: 160 °C
MDL Number: MFCD00464221
CAS Number: 14441-90-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76930d75a5a43a111a28540604eca0c495d181e6&vid=76930d75a5a43a111a28540604eca0c495d181e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR