5-Phenylbenzoxazole-2-thiol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Mercapto-5-phenylbenzoxazole, 5-Phenylbenzoxazoline-2(3H)-thione

P2011-1G P2011-5G
TCP2011-1GEA 115.66 CAD
TCP2011-1G TCP2011-5G
5-Phenylbenzoxazole-2-thiol ≥98.0% (by GC, titration analysis)
5-Phenylbenzoxazole-2-thiol

CAS Number: 17371-99-2
MDL Number: MFCD00022847
Molecular Formula: C13H9NOS
Molecular Weight: 227.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 213


Product Categories: Synthetic Organic Chemistry, Thiols
Other Product Names / Synonyms: 2-Mercapto-5-phenylbenzoxazole, 5-Phenylbenzoxazoline-2(3H)-thione

Formula: C₁₃H₉NOS
MW: 227.28 g/mol
Melting Pt: 213 °C
MDL Number: MFCD00022847
CAS Number: 17371-99-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5868a55a67b6775f9684fef05f33328702b0f2&vid=da5868a55a67b6775f9684fef05f33328702b0f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR