N-(2-Oxoethyl)phthalimide ≥98.0%

Supplier: TCI America

Synonyms: N-(Formylmethyl)phthalimide, Phthalimidoacetaldehyde

P2010-1G P2010-5G
TCP2010-1GEA 104.28 CAD
TCP2010-1G TCP2010-5G
N-(2-Oxoethyl)phthalimide ≥98.0%
N-(2-Oxoethyl)phthalimide

CAS Number: 2913-97-5
MDL Number: MFCD00023080
Molecular Formula: C10H7NO3
Molecular Weight: 189.17
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 114
Storage Temperature: <0°C


Other Product Names / Synonyms: N-(Formylmethyl)phthalimide

Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
Melting Pt: 114 °C
MDL Number: MFCD00023080
CAS Number: 2913-97-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f22ff30762a2ecd3c7d6c6c901459ff1d64782ed&vid=f22ff30762a2ecd3c7d6c6c901459ff1d64782ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR