2-Phenylethyl isovalerate ≥98.0%

Supplier: TCI America

Synonyms: Phenethyl 3-methylbutanoate, Phenethyl Isovalerate, Isovaleric Acid 2-Phenylethyl Ester, Isovaleric Acid Phenethyl Ester

P2009-25ML P2009-500ML
TCP2009-25MLEA 47.4 CAD
TCP2009-25ML TCP2009-500ML
2-Phenylethyl isovalerate ≥98.0%
2-Phenylethyl isovalerate

CAS Number: 140-26-1
MDL Number: MFCD00044322
Molecular Formula: C13H18O2
Molecular Weight: 206.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 268
Flash Point (°C): 100
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: Isovaleric Acid 2-Phenylethyl Ester, Phenethyl Isovalerate, Isovaleric Acid Phenethyl Ester

Formula: C₁₃H₁₈O₂
MW: 206.29 g/mol
Boiling Pt: 268 °C
Density: 0.98 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00044322
CAS Number: 140-26-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d39dfefa423b0a9535e773e5485d3019773d43a&vid=1d39dfefa423b0a9535e773e5485d3019773d43a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR