2-Phenylethyl isobutyrate ≥98.0%
Supplier: TCI America
Synonyms:
Phenethyl Isobutyrate, Isobutyric Acid Phenethyl Ester, Isobutyric Acid 2-Phenylethyl Ester, Phenethyl isobutyrate
CAS Number: 103-48-0
MDL Number: MFCD00026441
Molecular Formula: C12H16O2
Molecular Weight: 192.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 250
Flash Point (°C): 100
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: Isobutyric Acid 2-Phenylethyl Ester, Phenethyl Isobutyrate, Isobutyric Acid Phenethyl Ester
Formula:
C₁₂H₁₆O₂ Boiling Pt: 250 °C Density: 0.99 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00026441 CAS Number: 103-48-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...