2-Phenylethyl cinnamate ≥99.0%

Supplier: TCI America

Synonyms: Phenethyl cinnamate, Cinnamic Acid 2-Phenylethyl Ester, Phenethyl Cinnamate, Cinnamic Acid Phenethyl Ester

P2007-25G P2007-500G
TCP2007-25GEA 39.5 CAD
TCP2007-25G TCP2007-500G
2-Phenylethyl cinnamate ≥99.0%
2-Phenylethyl cinnamate

CAS Number: 103-53-7
MDL Number: MFCD00022050
Molecular Formula: C17H16O2
Molecular Weight: 252.31
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 57
Flash Point (°C): 113


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: Cinnamic Acid 2-Phenylethyl Ester, Phenethyl Cinnamate, Cinnamic Acid Phenethyl Ester

Formula: C₁₇H₁₆O₂
MW: 252.31 g/mol
Melting Pt: 57 °C
Flash Pt: 113 °C
MDL Number: MFCD00022050
CAS Number: 103-53-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e494a5b0536158849d997a08c6040fe6ce7f89d3&vid=e494a5b0536158849d997a08c6040fe6ce7f89d3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR