3-Pyridinecarboxamide oxime ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
P2003-1G
P2003-5G
TCP2003-1GEA
71.1
CAD
TCP2003-1G
TCP2003-5G
3-Pyridinecarboxamide oxime ≥98.0% (by HPLC, titration analysis)
3-Pyridinecarboxamide oxime
CAS Number: 1594-58-7
MDL Number: MFCD00265955
Molecular Formula: C6H7N3O
Molecular Weight: 137.14
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 132
Other Product Names / Synonyms: 3-Pyridylamidoxime, Nicotinamide Oxime
Formula:
C₆H₇N₃O Melting Pt: 132 °C |
MDL Number:
MFCD00265955 CAS Number: 1594-58-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...