9-Phenylcarbazole-3-boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

P2001-25G P2001-5G
TCP2001-25GEA 900.92 CAD
TCP2001-25G TCP2001-5G
9-Phenylcarbazole-3-boronic acid (contains varying amounts of Anhydride)
9-Phenylcarbazole-3-boronic acid

CAS Number: 854952-58-2
MDL Number: MFCD12196936
Molecular Formula: C18H14BNO2
Molecular Weight: 287.13
Form: Crystal
Color: White

Formula: C₁₈H₁₄BNO₂
MDL Number: MFCD12196936
CAS Number: 854952-58-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78847552d360ea46acbd88a12ef93517385528de&vid=78847552d360ea46acbd88a12ef93517385528de&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR