6(5H)-Phenanthridinone ≥98.0%

Supplier: TCI America

P1998-1G P1998-5G
TCP1998-1GEA 50.56 CAD
TCP1998-1G TCP1998-5G
6(5H)-Phenanthridinone ≥98.0%
6(5H)-Phenanthridinone

CAS Number: 1015-89-0
MDL Number: MFCD00004988
Molecular Formula: C13H9NO
Molecular Weight: 195.22
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 295
Lambda max.: 338 nm (EtOH)

Formula: C₁₃H₉NO
MW: 195.22
Melting Pt: 295 °C
MDL Number: MFCD00004988
CAS Number: 1015-89-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0189e62b1daf648338a044ad8787fa3412c0c07&vid=d0189e62b1daf648338a044ad8787fa3412c0c07&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR