4-(4-Phenylbutoxy)benzoic acid ≥98.0%
Supplier: TCI America
P1997-25G
P1997-5G
TCP1997-25GEA
206.66
CAD
TCP1997-25G
TCP1997-5G
4-(4-Phenylbutoxy)benzoic acid ≥98.0%
4-(4-Phenylbutoxy)benzoic acid
CAS Number: 30131-16-9
MDL Number: MFCD07787608
Molecular Formula: C17H18O3
Molecular Weight: 270.33
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 131
Formula:
C₁₇H₁₈O₃ MW: 270.33 g/mol Melting Pt: 131 °C |
MDL Number:
MFCD07787608 CAS Number: 30131-16-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...