1H-Pyrazole-4-carboxylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
P1978-1G
P1978-5G
TCP1978-1GEA
131.14
CAD
TCP1978-1G
TCP1978-5G
1H-Pyrazole-4-carboxylic acid ≥98.0% (by titrimetric analysis)
1H-Pyrazole-4-carboxylic acid
CAS Number: 37718-11-9
MDL Number: MFCD00077436
Molecular Formula: C4H4N2O2
Molecular Weight: 112.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 275
Formula:
C₄H₄N₂O₂ Melting Pt: 275 °C |
MDL Number:
MFCD00011558 CAS Number: 37718-11-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...