4-Phenoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: (4-Phenoxyphenyl)boronic acid, 4-Phenoxybenzeneboronic acid

P1974-25G P1974-5G
TCP1974-25GEA 279.34 CAD
TCP1974-25G TCP1974-5G
4-Phenoxyphenylboronic acid (contains varying amounts of Anhydride)
4-Phenoxyphenylboronic acid

CAS Number: 51067-38-0
MDL Number: MFCD00093312
Molecular Formula: C12H11BO3
Molecular Weight: 214.03
Form: Crystal
Color: White
Melting point (°C): 115


Other Product Names / Synonyms: 4-Phenoxybenzeneboronic Acid

Formula: C₁₂H₁₁BO₃
Melting Pt: 115 °C
MDL Number: MFCD00093312
CAS Number: 51067-38-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43e97b28e7800b18a4ae69ad884782cb3005cd02&vid=43e97b28e7800b18a4ae69ad884782cb3005cd02&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR