1,10-Phenanthroline-5,6-dione ≥98.0%
Supplier: TCI America
P1973-1G
P1973-5G
TCP1973-1GEA
247.41
CAD
TCP1973-1G
TCP1973-5G
1,10-Phenanthroline-5,6-dione ≥98.0%
1,10-Phenanthroline-5,6-dione
CAS Number: 27318-90-7
MDL Number: MFCD00014473
Molecular Formula: C12H6N2O2
Molecular Weight: 210.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 257
Lambda max.: 251 nm (MeOH)
Formula:
C₁₂H₆N₂O₂ MW: 210.19 g/mol Melting Pt: 257 °C |
MDL Number:
MFCD00014473 CAS Number: 27318-90-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...