3-(4-Pyridyl)acrylic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

P1972-1G P1972-5G
TCP1972-1GEA 97.96 CAD
TCP1972-1G TCP1972-5G
3-(4-Pyridyl)acrylic acid ≥97.0% (by GC, titration analysis)
3-(4-Pyridyl)acrylic acid

CAS Number: 5337-79-1
MDL Number: MFCD00040746
Molecular Formula: C8H7NO2
Molecular Weight: 149.15
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 275


Other Product Names / Synonyms: 4-Pyridineacrylic Acid

Formula: C₈H₇NO₂
Melting Pt: 275 °C
MDL Number: MFCD00040746
CAS Number: 5337-79-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c809bd09307ab0ae48ba7579a14a775aa804c030&vid=c809bd09307ab0ae48ba7579a14a775aa804c030&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR