Perphenazine ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

P1970-25G P1970-5G
TCP1970-25GEA 273.34 CAD
TCP1970-25G TCP1970-5G
Perphenazine ≥97.0% (by HPLC, titration analysis)
Perphenazine

CAS Number: 58-39-9
MDL Number: MFCD00056798
Molecular Formula: C21H26ClN3OS
Molecular Weight: 403.97
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 97
Storage Temperature: 0-10°C


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 2-Chloro-10-[3-[4-(2-hydroxyethyl)piperazin-1-yl]propyl]phenothiazine, 4-[3-(2-Chlorophenothiazin-10-yl)propyl]-1-piperazineethanol

Formula: C₂₁H₂₆ClN₃OS
Melting Pt: 97 °C
MDL Number: MFCD00056798
CAS Number: 58-39-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23983e07dcedfed11590b52c6c851923d8ce5718&vid=23983e07dcedfed11590b52c6c851923d8ce5718&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR