Phloretin ≥98.0% (by HPLC)

Supplier: TCI America

P1966-1G P1966-5G
TCP1966-1GEA 36.34 CAD
TCP1966-1G TCP1966-5G
Phloretin ≥98.0% (by HPLC)
Phloretin

CAS Number: 60-82-2
MDL Number: MFCD00002288
Molecular Formula: C15H14O5
Molecular Weight: 274.27
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 262
Storage Temperature: 0-10°C


Product Categories: Bioscience, Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 3-(4-Hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)-1-propanone, 2',4',6'-Trihydroxy-3-(4-hydroxyphenyl)propiophenone

Formula: C₁₅H₁₄O₅
Melting Pt: 262 °C
MDL Number: MFCD00002288
CAS Number: 60-82-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d7df91e4b3b7f11333a7e2148e7b2ab04512002&vid=3d7df91e4b3b7f11333a7e2148e7b2ab04512002&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR