3-Phenyl-3-(trifluoromethyl)-3H-diazirine ≥98.0% (by HPLC)
Supplier: TCI America
P1963-1G
P1963-200MG
TCP1963-1GEA
945.47
CAD
TCP1963-1G
TCP1963-200MG
3-Phenyl-3-(trifluoromethyl)-3H-diazirine ≥98.0% (by HPLC)
3-Phenyl-3-(trifluoromethyl)-3H-diazirine
CAS Number: 73899-14-6
Molecular Formula: C8H5F3N2
Molecular Weight: 186.14
Purity/Analysis Method: >98.0% (HPLC)
Form: Clear Liquid
Color: Pale Yellow
Storage Temperature: 0-10°C
Formula:
C₈H₅F₃N₂ MW: 186.14 g/mol |
MDL Number:
MFCD22041649 CAS Number: 73899-14-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...