Procaine ≥98.0%

Supplier: TCI America

Synonyms: 2-(Diethylamino)ethyl 4-Aminobenzoate, 4-Aminobenzoic acid 2-(Diethylamino)ethyl ester, 2-(Diethylamino)ethyl 4-aminobenzoate

P1961-25G P1961-5G
TCP1961-25GEA 231.47 CAD
TCP1961-25G TCP1961-5G
Procaine ≥98.0%
Procaine

CAS Number: 59-46-1
MDL Number: MFCD00007893
Molecular Formula: C13H20N2O2
Molecular Weight: 236.32
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 62


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 4-Aminobenzoic Acid 2-(Diethylamino)ethyl Ester, 2-(Diethylamino)ethyl 4-Aminobenzoate

Formula: C₁₃H₂₀N₂O₂
Melting Pt: 62 °C
MDL Number: MFCD00007893
CAS Number: 59-46-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adeb0588ab34d88c2207af628b7b955f21b62b29&vid=adeb0588ab34d88c2207af628b7b955f21b62b29&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR