Methyl palmitoleate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Palmitoleic acid methyl ester, (Z)-Methyl hexadec-9-enoate

P1958-100MG P1958-1G
TCP1958-100MGEA 66.36 CAD
TCP1958-100MG TCP1958-1G
Methyl palmitoleate ≥98.0% (by GC)
Methyl palmitoleate

CAS Number: 1120-25-8
MDL Number: MFCD00042911
Molecular Formula: C17H32O2
Molecular Weight: 268.44
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 126
Flash Point (°C): 113
Specific Gravity (20/20): 0.88
Storage Temperature: <0°C


Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: cis-9-Hexadecenoic Acid Methyl Ester, Methyl Palmitoleate, Palmitoleic Acid Methyl Ester

Formula: C₁₇H₃₂O₂
MW: 268.44 g/mol
Boiling Pt: 126 °C
Density: 0.88 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00042911
CAS Number: 1120-25-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76275f2a36daf689277b56b2b9f92e086bb9feb6&vid=76275f2a36daf689277b56b2b9f92e086bb9feb6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR