2-Phenoxyethanol ≥98.5% (by GC) for biochemical research

Supplier: TCI America

Synonyms: ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether

P1953-25G P1953-5G
TCP1953-25GEA 85.32 CAD
TCP1953-25G TCP1953-5G
2-Phenoxyethanol ≥98.5% (by GC) for biochemical research
2-Phenoxyethanol

[for Biochemical Research]
CAS Number: 122-99-6
MDL Number: MFCD00002857
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 237
Flash Point (°C): 127
Freezing point (°C): 12
Specific Gravity (20/20): 1.11


Product Categories: Bioscience, Preservatives for Biochemical Research
Other Product Names / Synonyms: Ethylene Glycol Monophenyl Ether, Phenyl Cellosolve, Phenyl Glycol

Formula: C₆H₅OCH₂CH₂OH
MW: 138.17 g/mol
Boiling Pt: 245
Melting Pt: 13
Density: 1.11 (20 °C)
Flash Pt: 127 °C
MDL Number: MFCD00002857
CAS Number: 122-99-6
Merck Index: 12,07410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edcdaa94999c3ec8e43c85749e3dec570a8bb1df&vid=abd0e27e965c30e9df80e4f69022d8eaf8c16347&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR