1-Phenyl-3-(2,4,6-trimethylphenyl)-2-pyrazoline ≥98.0% (by GC)

Supplier: TCI America

P1951-1G P1951-5G
TCP1951-1GEA 235.42 CAD
TCP1951-1G TCP1951-5G
1-Phenyl-3-(2,4,6-trimethylphenyl)-2-pyrazoline ≥98.0% (by GC)
1-Phenyl-3-(2,4,6-trimethylphenyl)-2-pyrazoline

CAS Number: 60078-97-9
Molecular Formula: C18H20N2
Molecular Weight: 264.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 97


Other Product Names / Synonyms: 3-Mesityl-1-phenyl-2-pyrazoline

Formula: C₁₈H₂₀N₂
Melting Pt: 97 °C
MDL Number: MFCD00153039
CAS Number: 60078-97-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c1d214a1829405870d6dbcfbf5d39513d4c9c7d&vid=6c1d214a1829405870d6dbcfbf5d39513d4c9c7d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR