Polyethylene glycolbis(3-aminopropyl)ether ≥95.0% (by titrimetric analysis)
Supplier: TCI America
P1948-1G
P1948-5G
TCP1948-1GEA
64.46
CAD
TCP1948-1G
TCP1948-5G
Polyethylene glycolbis(3-aminopropyl)ether ≥95.0% (by titrimetric analysis)
Polyethylene glycolbis(3-aminopropyl)ether
CAS Number: 34901-14-9
MDL Number: MFCD00197739
Form: Crystal
Color: White
Melting point (°C): 54
Melting Pt:
54 °C |
MDL Number:
MFCD00197739 CAS Number: 34901-14-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...