(4S,4'S)-2,2'-(Pentane-3,3'-diyl)bis(4-benzyl-4,5-dihydrooxazole) ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P1947-1G P1947-200MG
TCP1947-1GEA 725.69 CAD
TCP1947-1G TCP1947-200MG
(4S,4'S)-2,2'-(Pentane-3,3'-diyl)bis(4-benzyl-4,5-dihydrooxazole) ≥98.0% (by titrimetric analysis)
(4S,4'S)-2,2'-(Pentane-3,3'-diyl)bis(4-benzyl-4,5-dihydrooxazole)

CAS Number: 160191-64-0
Molecular Formula: C25H30N2O2
Molecular Weight: 390.53
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Pale Yellow
Specific rotation [a]20/D: -21 deg (C=2.5, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (S,S)-3,3-Bis(4-benzyl-2-oxazolin-2-yl)pentane, (S,S)-2,2'-(Diethylmethylene)bis(4-benzyl-2-oxazoline)

Formula: C₂₅H₃₀N₂O₂
MDL Number: MFCD20265601
CAS Number: 160191-64-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f38204027aa2f419872645427b58446b3767942d&vid=f38204027aa2f419872645427b58446b3767942d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR