1-Propylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

P1942-1G P1942-25G P1942-5G
TCP1942-1GEA 39.5 CAD
TCP1942-1G TCP1942-25G TCP1942-5G
1-Propylboronic acid (contains varying amounts of Anhydride)
1-Propylboronic acid

CAS Number: 17745-45-8
MDL Number: MFCD01074564
Molecular Formula: C3H9BO2
Molecular Weight: 87.91
Form: Crystal
Color: White
Melting point (°C): 106


Other Product Names / Synonyms: 1-Propaneboronic Acid

Formula: C₃H₉BO₂
Melting Pt: 106 °C
MDL Number: MFCD01074564
CAS Number: 17745-45-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=635f86565ea23f8866a68dbe037f567e3d39e00c&vid=635f86565ea23f8866a68dbe037f567e3d39e00c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR