(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol ≥98.0% (by GC)

Supplier: TCI America

P1934-25G P1934-5G
TCP1934-25GEA 278.08 CAD
TCP1934-25G TCP1934-5G
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol ≥98.0% (by GC)
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol

CAS Number: 18680-27-8
MDL Number: MFCD00077851
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 57
Specific rotation [a]20/D: 9 deg (C=6.5, Toluene)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Bicyclic Monoterpenes
Other Product Names / Synonyms: (1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol

Formula: C10H18O2
Melting Pt: 57 °C
MDL Number: MFCD00077851
CAS Number: 18680-27-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d51d330e4b9fc95ec71c1cebe467ef0d62abf534&vid=d51d330e4b9fc95ec71c1cebe467ef0d62abf534&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR