4-(1-Pyrrolidinyl)piperidine ≥98.0%
Supplier: TCI America
P1932-25G
P1932-5G
TCP1932-25GEA
728.3
CAD
TCP1932-25G
TCP1932-5G
4-(1-Pyrrolidinyl)piperidine ≥98.0%
4-(1-Pyrrolidinyl)piperidine
CAS Number: 5004-07-9
MDL Number: MFCD00038011
Molecular Formula: C9H18N2
Molecular Weight: 154.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 60
Formula:
C₉H₁₈N₂ MW: 154.2526 g/mol Melting Pt: 60 °C |
MDL Number:
MFCD00038011 CAS Number: 5004-07-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...