Pentyl valerate ≥98.0% (by GC)

Supplier: TCI America

P1929-25ML P1929-500ML
TCP1929-25MLEA 42.66 CAD
TCP1929-25ML TCP1929-500ML
Pentyl valerate ≥98.0% (by GC)
Pentyl valerate

CAS Number: 2173-56-0
MDL Number: MFCD00042904
Molecular Formula: C10H20O2
Molecular Weight: 172.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 205
Flash Point (°C): 81
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Valeric Acid Pentyl Ester, Pentyl Pentanoate, Pentanoic Acid Pentyl Ester, Amyl Pentanoate, Amyl Valerate

Formula: C₁₀H₂₀O₂
Boiling Pt: 205 °C
Density: 0.87 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00042904
CAS Number: 2173-56-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc74710865fa5719f9f0e213cac628bbbb79e49f&vid=cc74710865fa5719f9f0e213cac628bbbb79e49f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR