Pterostilbene ≥98.0% (by HPLC)

Supplier: TCI America

P1924-100MG P1924-1G
TCP1924-100MGEA 28.93 CAD
TCP1924-100MG TCP1924-1G
Pterostilbene ≥98.0% (by HPLC)
Pterostilbene

CAS Number: 537-42-8
MDL Number: MFCD00238710
Molecular Formula: C16H16O3
Molecular Weight: 256.30
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Lambda max.: 321 nm (MeOH)Storage Temperature: 0-10°C


Product Categories: Bioscience, Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 4'-Hydroxy-3,5-dimethoxy-trans-stilbene, trans-1-(3,5-Dimethoxyphenyl)-2-(4-hydroxyphenyl)ethylene

Formula: C₁₆H₁₆O₃
MDL Number: MFCD00238710
CAS Number: 537-42-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=189a64bef8a48fcf3b288882994e128ba4448413&vid=189a64bef8a48fcf3b288882994e128ba4448413&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR