Polyethyleneimine ∼30% in water for biochemical research

Supplier: TCI America

Synonyms: PolyaziridinePolyethyleniminePolyethylenimine (branched)Polyaziridine Mw 25000PEICorcatPolymin PEthyleneimine PolymerAziridinePoly(ethyleneimine)

P1921-100G P1921-25G
TCP1921-100GEA 76.88 CAD
TCP1921-100G TCP1921-25G
Polyethyleneimine ∼30% in water for biochemical research
Polyethyleneimine

[for Biochemical Research]
CAS Number: 9002-98-6
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: Ethyleneimine Polymer

Formula: (CH₂CH₂NH)n
MDL Number: MFCD00084427
CAS Number: 9002-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR