5-Azaindole ≥98.0% (by GC, titration analysis)
Supplier: TCI America
P1896-100MG
P1896-1G
TCP1896-100MGEA
31.6
CAD
TCP1896-100MG
TCP1896-1G
5-Azaindole ≥98.0% (by GC, titration analysis)
5-Azaindole
CAS Number: 271-34-1
MDL Number: MFCD00955936
Molecular Formula: C7H6N2
Molecular Weight: 118.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 132
Melting point (°C): 113
Lambda max.: 264 nm (H2O)
Other Product Names / Synonyms: 5-Azaindole
Formula:
C₇H₆N₂ Boiling Pt: 132 °C Melting Pt: 113 °C |
MDL Number:
MFCD00955936 CAS Number: 271-34-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...