Puerarin ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 8-(beta-D-Glucopyranosyl)-4',7-dihydroxyisoflavone

P1886-1G P1886-200MG
TCP1886-1GEA 79 CAD
TCP1886-1G TCP1886-200MG
Puerarin ≥98.0% (by HPLC, titration analysis)
Puerarin

CAS Number: 3681-99-0
MDL Number: MFCD00076007
Molecular Formula: C21H20O9
Molecular Weight: 416.38
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 187
Lambda max.: 303 nm (MeOH)Storage Temperature: 0-10°C


Product Categories: Bioscience, Glycosides, Glucose, Crude Medicine Ingredients for Research and Experimental Use, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 8-(beta-D-Glucopyranosyl)-4',7-dihydroxyisoflavone

Formula: C₂₁H₂₀O₉
MW: 416.38 g/mol
Melting Pt: 187 °C
MDL Number: MFCD00076007
CAS Number: 3681-99-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca08dfa166a7bb2e9b8186adef69a9eab7f93410&vid=51a01c7b009a36a9c74fb6fd9a5f42a13cf534d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR