4-Propylcyclohexanol (cis- and trans- mixture) ≥98.0% (by GC)
Supplier: TCI America
P1874-1G
P1874-5G
TCP1874-1GEA
158
CAD
TCP1874-1G
TCP1874-5G
4-Propylcyclohexanol (cis- and trans- mixture) ≥98.0% (by GC)
4-Propylcyclohexanol (cis- and trans- mixture)
CAS Number: 52204-65-6
MDL Number: MFCD00091618
Molecular Formula: C9H18O
Molecular Weight: 142.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.91
Formula:
C₉H₁₈O Density: 0.91 (20 °C) |
MDL Number:
MFCD00091618 CAS Number: 52204-65-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...