5-Methylphenazinium methyl sulfate ≥98.0% (by HPLC, total nitrogen) for biochemical research

Supplier: TCI America

P1872-1G P1872-5G
TCP1872-1GEA 66.36 CAD
TCP1872-1G TCP1872-5G
5-Methylphenazinium methyl sulfate ≥98.0% (by HPLC, total nitrogen) for biochemical research
5-Methylphenazinium methyl sulphate

[for Biochemical Research]
CAS Number: 299-11-6
MDL Number: MFCD00011923
Molecular Formula: C14H14N2O4S
Molecular Weight: 306.34
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: Yellow
Melting point (°C): 160
Storage Temperature: 0-10°C


Product Categories: Bioscience, Redox Research Reagents
Other Product Names / Synonyms: N-Methylphenazonium Methosulfate, N-Methylphenazonium Methyl Sulfate, Phenazine Methosulfate

Formula: C₁₄H₁₄N₂O₄S
Melting Pt: 160 °C
MDL Number: MFCD00011923
CAS Number: 299-11-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22dfd45999ff362c6592f1cc0a0720851722b483&vid=5bb413d32272a7942ba921d8fa10d4075af963df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR