Palladium(II) trifluoroacetate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P1870-1G P1870-5G
TCP1870-1GEA 562.48 CAD
TCP1870-1G TCP1870-5G
Palladium(II) trifluoroacetate ≥98.0% (by titrimetric analysis)
Palladium (II) trifluoroacetate

CAS Number: 42196-31-6
MDL Number: MFCD00013204
Molecular Formula: C4F6O4Pd
Molecular Weight: 332.45
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 220


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Oxidation
Other Product Names / Synonyms: Trifluoroacetic Acid Palladium(II) Salt

Formula: C₄F₆O₄Pd
Melting Pt: 220 °C
MDL Number: MFCD00013204
CAS Number: 42196-31-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fbd18f5c5502d512a16976f063ec093a4ed706c&vid=5fbd18f5c5502d512a16976f063ec093a4ed706c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR