1-Phenyl-1H-pyrazole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Phenyl-1H-pyrazole, 1-Phenylpyrazole

P1868-25G P1868-5G
TCP1868-25GEA 232.26 CAD
TCP1868-25G TCP1868-5G
1-Phenyl-1H-pyrazole ≥98.0% (by GC)
1-Phenyl-1H-pyrazole

CAS Number: 1126-00-7
MDL Number: MFCD00003112
Molecular Formula: C9H8N2
Molecular Weight: 144.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Slightly Pale Yellow
Boiling point (°C): 142
Melting point (°C): 11
Flash Point (°C): 113
Specific Gravity (20/20): 1.11
Lambda max.: 255 nm (EtOH)

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 142 °C
Melting Pt: 11 °C
Density: 1.11 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00003112
CAS Number: 1126-00-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c91308f2849e1a5fb2fca61dacf41b09374d6187&vid=c91308f2849e1a5fb2fca61dacf41b09374d6187&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR