2-Piperazinone ≥98.0% (by GC)
Supplier: TCI America
P1867-1G
P1867-5G
TCP1867-1GEA
37.92
CAD
TCP1867-1G
TCP1867-5G
2-Piperazinone ≥98.0% (by GC)
2-Piperazinone
CAS Number: 5625-67-2
MDL Number: MFCD01318687
Molecular Formula: C4H8N2O
Molecular Weight: 100.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Slightly Pale Yellow
Boiling point (°C): 164
Melting point (°C): 134
Other Product Names / Synonyms: Piperazin-2-one, 2-Oxopiperazine
Formula:
C₄H₈N₂O Boiling Pt: 164 °C Melting Pt: 134 °C |
MDL Number:
MFCD01318687 CAS Number: 5625-67-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...