1H-Pyrazole-3-carboxylic acid ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 3-Pyrazolecarboxylic acid, Pyrazole-3-carboxylic acid

P1862-1G P1862-5G
TCP1862-1GEA 58.46 CAD
TCP1862-1G TCP1862-5G
1H-Pyrazole-3-carboxylic acid ≥98.0% (by HPLC)
1H-Pyrazole-3-carboxylic acid

CAS Number: 1621-91-6
MDL Number: MFCD00077436
Molecular Formula: C4H4N2O2
Molecular Weight: 112.09
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 213
Lambda max.: 260 nm (MeOH)

Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Melting Pt: 213 °C
MDL Number: MFCD00077436
CAS Number: 1621-91-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf3d0dbad8d914b4e192fa84820d774ea96ada35&vid=bf3d0dbad8d914b4e192fa84820d774ea96ada35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR