3-(1-Pyrrolylmethyl)pyridine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

P1853-1G P1853-5G
TCP1853-1GEA 211.72 CAD
TCP1853-1G TCP1853-5G
3-(1-Pyrrolylmethyl)pyridine ≥97.0% (by GC, titration analysis)
3-(1-Pyrrolylmethyl)pyridine

CAS Number: 80866-95-1
MDL Number: MFCD00006405
Molecular Formula: C10H10N2
Molecular Weight: 158.20
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 60

Formula: C₁₀H₁₀N₂
Melting Pt: 60 °C
MDL Number: MFCD00006405
CAS Number: 80866-95-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=029788db9ba97cbb31614d2a619566e7d31b51cc&vid=029788db9ba97cbb31614d2a619566e7d31b51cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR