Potassium cyclopropyltrifluoroborate ≥97.0%

Supplier: TCI America

Synonyms: Cyclopropyltrifluoroboric acid potassium salt

P1846-1G P1846-5G
TCP1846-1GEA 109.02 CAD
TCP1846-1G TCP1846-5G
Potassium cyclopropyltrifluoroborate ≥97.0%
Potassium cyclopropyltrifluoroborate

CAS Number: 1065010-87-8
MDL Number: MFCD09265031
Molecular Formula: C3H5BF3K
Molecular Weight: 147.98
Purity/Analysis Method: >97.0% (W)
Form: Crystal
Color: White

Formula: C₃H₅BF₃K
MDL Number: MFCD09265031
CAS Number: 1065010-87-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0fe5148068e20df021639fd459fd5112a2cb82a&vid=e0fe5148068e20df021639fd459fd5112a2cb82a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (W)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR